Assay Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026163.2 | NP_080439.1 | 8 - 9 | 1747 | 63 | |
| GenBank mRNA | AK004650.1 | - | 8 - 9 | 1748 | 63 | |
| AK005020.1 | - | 8 - 9 | 1779 | 63 | ||
| AK049481.1 | - | 8 - 9 | 1748 | 63 | ||
| AK167660.1 | - | 8 - 9 | 1749 | 63 | ||
| BC018487.1 | - | 2 - 3 | 303 | 63 | ||
| BC037133.1 | - | 3 - 4 | 353 | 63 | ||
| BC068279.1 | - | 6 - 7 | 1027 | 63 | ||
| BC138252.1 | - | 8 - 9 | 1747 | 8861271 | 63 | |
| BC138253.1 | - | 8 - 9 | 1747 | 8861272 | 63 | |
More Information
Set Membership: |
|

Pkp2
Genomic Map