Assay Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285805.1 | NP_001272734.1 | 5 - 6 | 282 | 76 | |
| NM_019778.2 | NP_062752.2 | 3 - 4 | 229 | 76 | ||
| NM_181058.1 | NP_851401.1 | 3 - 4 | 229 | 76 | ||
| GenBank mRNA | AK005028.2 | - | 3 - 4 | 230 | 76 | |
| AK052686.1 | - | 2 - 3 | 556 | 76 | ||
| AK078664.1 | - | 213 | 76 | |||
| BC031114.1 | - | 3 - 4 | 173 | 4971586 | 76 | |
| BC056446.1 | - | 5 - 6 | 254 | 5703319 | 76 | |
| BI659024.1 | - | 7 - 8 | 435 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005028.2, BC031114.1, BC056446.1, NM_001285805.1, NM_019778.2, NM_181058.1 transcripts.
Set Membership: |
|

Zbtb20
Genomic Map