| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600013475.mm.1 |
NM_001159329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013475.mm.1 |
NM_001159329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013475.mm.1 |
NM_025332.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013475.mm.1 |
NM_025332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013475.mm.1 |
TC1600001648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013476.mm.1 |
NM_001159329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013476.mm.1 |
NM_001159329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013476.mm.1 |
NM_025332.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013476.mm.1 |
NM_025332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013476.mm.1 |
TC1600001648.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013480.mm.1 |
NM_001159329.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013480.mm.1 |
NM_001159329.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013480.mm.1 |
NM_025332.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013480.mm.1 |
NM_025332.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013480.mm.1 |
TC1600001648.mm.1 |
MTA TC |