Assay Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024273.2 | NP_077235.2 | 6 - 7 | 839 | 84 | |
| GenBank mRNA | AK081781.1 | - | 6 - 7 | 782 | 84 | |
| BC002243.1 | - | 6 - 7 | 801 | 3492342 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686478 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686477 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686476 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2429714 | BC002243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2429714 | NM_024273.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2429714 | AK012774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2429714 | AK081781.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1975095 | BC002243.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039463 | NM_175523.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039463 | AK034199.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039463 | NM_001122667.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039463 | NM_153588.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039463 | AK020592.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117529 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544924 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411312 | AK041649.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150225 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295132 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 629239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230272 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419747 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 955818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52698 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 326197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150022 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 113413 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162448 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123292 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039463 | NM_175523.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039463 | AK034199.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039463 | NM_001122667.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039463 | NM_153588.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039463 | AK020592.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2429714 | BC002243.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2429714 | NM_024273.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2429714 | AK012774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2429714 | AK081781.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686465 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686493 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686489 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686480 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686482 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686486 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686462 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30686478 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162448 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08569360 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123292 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08569344 | - | - |
| Whole Mouse Genome G4122A | A_51_P468770 | BC002243.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P483885 | AK078465.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P139871 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123292 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162448 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12632615 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12632610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12632612 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12632627 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12632625 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686477 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686472 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686473 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686475 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686476 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30686478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30686339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686467 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686359 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686418 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12667225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12655341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12997167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686356 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686480 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12827484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686347 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686438 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686422 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686354 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13011980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12560847 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686440 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13006065 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686425 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686413 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686349 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12701231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12655343 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12827485 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686478 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12632629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686401 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686495 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12560849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686470 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13011979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686489 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12893036 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686415 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686465 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30686460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10439464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439469 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439468 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439465 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420329_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10439463 | - | - |
| Mouse Genome 430 2.0 Array | 1420329_at | - | - |
| Mouse Genome 430 2.0 Array | 1420329_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76916_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76916_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600013134.mm.1 | NM_024273.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013134.mm.1 | NM_024273.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013134.mm.1 | TC1600001606.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600013136.mm.1 | NM_024273.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013136.mm.1 | NM_024273.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600013136.mm.1 | TC1600001606.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420329_at | - | - |
| Mouse Expression Set 430 | 1420329_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420329_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420329_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420329_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420329_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 582226 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2838965 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2838965 | - | - |

Timmdc1
Genomic Map