Assay Details
| Gene Symbol: Gm6815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102685.1 | 1 | 2907 | 163 | ||
| GenBank mRNA | AK133264.1 | - | 1 | 2909 | 163 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm6815
Genomic Map