Assay Details
| Gene Symbol: Ydjc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026940.4 | NP_081216.1 | 4 - 5 | 616 | 62 | |
| GenBank mRNA | AK007503.1 | - | 4 - 5 | 605 | 62 | |
| BC115584.1 | - | 4 - 5 | 604 | 40102346 | 62 | |
| BC115585.1 | - | 4 - 5 | 604 | 40102347 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
biological_process
molecular_function
catalytic activity
histone deacetylase activity
protein-N-terminal asparagine amidohydrolase activity
UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
iprodione amidohydrolase activity
(3,5-dichlorophenylurea)acetate amidohydrolase activity
4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
didemethylisoproturon amidohydrolase activity
N-isopropylacetanilide amidohydrolase activity
N-cyclohexylformamide amidohydrolase activity
isonicotinic acid hydrazide hydrolase activity
cis-aconitamide amidase activity
gamma-N-formylaminovinylacetate hydrolase activity
N2-acetyl-L-lysine deacetylase activity
O-succinylbenzoate synthase activity
indoleacetamide hydrolase activity
N-acetylcitrulline deacetylase activity
metal ion binding
N-acetylgalactosamine-6-phosphate deacetylase activity
diacetylchitobiose deacetylase activity
chitooligosaccharide deacetylase activity
carbohydrate metabolic process
biological_process
molecular_function
catalytic activity
histone deacetylase activity
protein-N-terminal asparagine amidohydrolase activity
UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
iprodione amidohydrolase activity
(3,5-dichlorophenylurea)acetate amidohydrolase activity
4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
didemethylisoproturon amidohydrolase activity
N-isopropylacetanilide amidohydrolase activity
N-cyclohexylformamide amidohydrolase activity
isonicotinic acid hydrazide hydrolase activity
cis-aconitamide amidase activity
gamma-N-formylaminovinylacetate hydrolase activity
N2-acetyl-L-lysine deacetylase activity
O-succinylbenzoate synthase activity
indoleacetamide hydrolase activity
N-acetylcitrulline deacetylase activity
metal ion binding
N-acetylgalactosamine-6-phosphate deacetylase activity
diacetylchitobiose deacetylase activity
chitooligosaccharide deacetylase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572229 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572231 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202244 | AK032396.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P202244 | AK032396.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P162298 | BC030425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P162298 | NM_026940.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P162298 | AK007503.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P162298 | BC030425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P162298 | NM_026940.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P162298 | AK007503.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174802 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202244 | AK032396.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P202244 | AK032396.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P162298 | BC030425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P162298 | NM_026940.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P162298 | AK007503.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P162298 | BC030425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P162298 | NM_026940.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P162298 | AK007503.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572196 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572217 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572210 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572215 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572219 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572198 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572193 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572208 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572206 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572202 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572203 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572204 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30572200 | - | - |
| Whole Mouse Genome G4122A | A_51_P202244 | AK032396.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202244 | AK032396.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162298 | BC030425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162298 | NM_026940.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162298 | AK007503.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162298 | BC030425.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162298 | NM_026940.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P162298 | AK007503.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12707933 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677753 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12488510 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12677751 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12769155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12707937 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12769156 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12488512 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12488515 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12707934 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12707935 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572231 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30572229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30572187 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572227 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12488512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572219 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572200 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572180 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12608202 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572222 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12608216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12529273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572210 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572198 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12529276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572186 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572196 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572178 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572190 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572181 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12529280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12608204 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12488510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12488515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572192 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12529272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572203 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30572220 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10433966 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433967 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433968 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433970 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433969 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10433965 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428550_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10433963 | - | - |
| Mouse Genome 430 2.0 Array | 1428550_at | - | - |
| Mouse Genome 430 2.0 Array | 1428550_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69101_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69101_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600001689.mm.1 | NM_026940.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001689.mm.1 | NM_026940.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001689.mm.1 | TC1600000208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001690.mm.1 | NM_026940.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001690.mm.1 | NM_026940.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001690.mm.1 | TC1600000208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600001692.mm.1 | NM_026940.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001692.mm.1 | NM_026940.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600001692.mm.1 | TC1600000208.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428550_at | - | - |
| Mouse Expression Set 430 | 1428550_PM_at | - | - |
| Murine Genome U74v2 Set | 165441_at | - | - |
| Murine Genome U74v2 Set | 165441_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428550_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428550_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2883952 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2883952 | - | - |

Ydjc
Genomic Map