Assay Details
| Gene Symbol: Rnf186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025786.3 | NP_080062.4 | 1 | 1206 | 93 | |
| GenBank mRNA | AK020617.1 | - | 1 | 1115 | 93 | |
| AK078919.1 | - | 1 | 1201 | 93 | ||
| BC011492.1 | - | 1 | 1171 | 4207836 | 93 | |
| CF379387.1 | - | 1 | 267 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159934 | BC011492.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159934 | CF379387.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159934 | NM_025786.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2159934 | AK078919.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 315730 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137705 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159934 | BC011492.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159934 | CF379387.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159934 | NM_025786.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2159934 | AK078919.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23348881 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137705 | - | - |
| Whole Mouse Genome G4122A | A_51_P272023 | BC011492.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272023 | CF379387.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272023 | NM_025786.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272023 | AK020617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P272023 | AK078919.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110476 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137705 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04046297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04046300 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03733127 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03733140 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04046298 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03733134 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03733137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23348930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03757858 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04085135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03790341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03520168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03855593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03733140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03757862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03757853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03424783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23348888 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509597 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509598 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509599 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424794_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509596 | - | - |
| Mouse Genome 430 2.0 Array | 1424794_at | - | - |
| Mouse Genome 430 2.0 Array | 1424794_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66825_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66825_at | - | - |
| Mouse Expression Set 430 | 1424794_PM_at | - | - |
| Mouse Expression Set 430 | 1424794_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424794_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424794_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424794_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424794_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 334943 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 520286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520286 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2663996 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2663996 | - | - |
| mouse-6 v1.1 (Illumina) | scl24781.1.87_255-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520286 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24781.1.87_255-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2663996 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2663996 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24781.1.87_255-S | - | - |

Rnf186
Genomic Map