Assay Details
| Gene Symbol: Ndufv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030087.2 | NP_084363.2 | 3 - 4 | 1365 | 86 | |
| GenBank mRNA | AK154191.1 | - | 3 - 4 | 1298 | 86 | |
| AK154477.1 | - | 3 - 4 | 1344 | 86 | ||
| BF318157.1 | - | 1 - 2 | 86 | |||
| BU755401.1 | - | 1 - 2 | 86 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31140250 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31140249 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | AK154477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | BC013640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | BF318157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | NM_030087.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | AK154191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | AK154477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | BC013640.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | BF318157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | NM_030087.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P140211 | AK154191.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2022634 | AK160289.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184180 | BF318157.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184180 | NM_001083891.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184180 | NM_030087.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184180 | AK077433.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 121684 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493298 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 794608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 784189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111133 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 16281 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84657 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151329 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147618 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | AK154477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | BC013640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | BF318157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | NM_030087.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | AK154191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | AK154477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | BC013640.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | BF318157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | NM_030087.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P140211 | AK154191.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184180 | BF318157.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184180 | NM_001083891.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184180 | NM_030087.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184180 | AK077433.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31140257 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151329 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147618 | - | - |
| Whole Mouse Genome G4122A | A_51_P140211 | AK154477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | BC013640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | BF318157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | NM_030087.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | AK154191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | AK154477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | BC013640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | BF318157.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | NM_030087.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P140211 | AK154191.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170054 | AK154477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170054 | AK160289.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170054 | BC013640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170054 | BU555486.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P170054 | BY064612.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175897 | AK154477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175897 | BC013640.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175897 | NM_030087.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P175897 | AK154191.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P100131 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151329 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147618 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13498941 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13498931 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31140249 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31140250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140301 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31140307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13556430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140262 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13514861 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13327206 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140377 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13530083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140273 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13556433 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13556424 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13094487 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13327207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140287 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13576183 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140292 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31140296 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443812 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443809 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443810 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443813 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424628_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443808 | - | - |
| Mouse Genome 430 2.0 Array | 1424628_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1424628_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1445864_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1445864_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78330_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 78330_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700004937.mm.1 | NM_001083891.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004937.mm.1 | NM_001083891.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004937.mm.1 | NM_030087.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004937.mm.1 | NM_030087.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700004937.mm.1 | TC1700000539.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424628_PM_a_at | - | - |
| Mouse Expression Set 430 | 1424628_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424628_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424628_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424628_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424628_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 706669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4050180 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4850026 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050180 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004850026 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688822 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2668977 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2668977 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688822 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001719.1_218-S | - | - |
| mouse-6 v1.1 (Illumina) | scl50873.6.1_76-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4850026 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050180 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001719.1_218-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50873.6.1_76-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4850026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050180 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688822 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2668977 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3008277 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3008277 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2668977 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688822 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001719.1_218-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50873.6.1_76-S | - | - |

Ndufv3
Genomic Map