| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500007581.mm.1 |
NM_181728.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007581.mm.1 |
TC0500000871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007588.mm.1 |
NM_181728.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007588.mm.1 |
TC0500000871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007583.mm.1 |
NM_181728.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007583.mm.1 |
TC0500000871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007585.mm.1 |
NM_181728.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007585.mm.1 |
TC0500000871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007586.mm.1 |
NM_181728.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007586.mm.1 |
TC0500000871.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500007582.mm.1 |
NM_181728.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500007582.mm.1 |
TC0500000871.mm.1 |
MTA TC |