Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 12 - 13 | 1695 | 80 | |
| GenBank mRNA | AI255825.1 | - | 3 - 4 | 80 | ||
| AJ011080.1 | - | 12 - 13 | 1723 | 80 | ||
| BC100597.1 | - | 12 - 13 | 1695 | 6767654 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afm
Genomic Map