Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 13 - 14 | 1818 | 95 | |
| GenBank mRNA | AI255825.1 | - | 2 - 3 | 95 | ||
| BC100597.1 | - | 13 - 14 | 1818 | 6767654 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afm
Genomic Map