Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 1 - 2 | 125 | 83 | |
| GenBank mRNA | AK143987.1 | - | 1 - 2 | 126 | 83 | |
| BC100597.1 | - | 1 - 2 | 125 | 6767654 | 83 | |
More Information
Set Membership: |
|

Afm
Genomic Map