Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 5 - 6 | 661 | 132 | |
| GenBank mRNA | AJ011080.1 | - | 5 - 6 | 689 | 132 | |
| AK143987.1 | - | 5 - 6 | 662 | 132 | ||
| BC026681.1 | - | 5 - 6 | 579 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Afm
Genomic Map