Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 6 - 7 | 756 | 113 | |
| GenBank mRNA | AJ011080.1 | - | 6 - 7 | 784 | 113 | |
| AK143987.1 | - | 6 - 7 | 757 | 113 | ||
| BC026681.1 | - | 6 - 7 | 674 | 113 | ||
| BC100597.1 | - | 6 - 7 | 756 | 6767654 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afm
Genomic Map