Assay Details
| Gene Symbol: Tff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009363.3 | NP_033389.2 | 1 - 2 | 114 | 90 | |
| GenBank mRNA | AK003093.1 | - | 1 - 2 | 115 | 90 | |
| BC050086.1 | - | 1 - 2 | 75 | 90 | ||
| BC087889.1 | - | 1 - 2 | 92 | 30316277 | 90 | |
| X51697.1 | - | 1 - 2 | 79 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tff2
Genomic Map