Assay Details
| Gene Symbol: Tff2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009363.3 | NP_033389.2 | 3 - 4 | 415 | 59 | |
| GenBank mRNA | AK003093.1 | - | 3 - 4 | 416 | 59 | |
| BC050086.1 | - | 3 - 4 | 376 | 59 | ||
| BC087889.1 | - | 3 - 4 | 393 | 30316277 | 59 | |
| X51697.1 | - | 3 - 4 | 380 | 59 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell proliferation
calcium-mediated signaling
regulation of cell migration
negative regulation of macrophage activation
negative regulation of inflammatory response
negative regulation of gastric acid secretion
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
CXCR4 chemokine receptor binding
positive regulation of cell proliferation
calcium-mediated signaling
regulation of cell migration
negative regulation of macrophage activation
negative regulation of inflammatory response
negative regulation of gastric acid secretion
chemokine-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
CXCR4 chemokine receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | NM_009363.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | AK003093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | BC050086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | BC087889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | X51697.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | NM_009363.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | AK003093.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | BC050086.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | BC087889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P480602 | X51697.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454438 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150359 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | NM_009363.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | AK003093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | BC050086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | BC087889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | X51697.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | NM_009363.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | AK003093.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | BC050086.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | BC087889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P480602 | X51697.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31138131 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150359 | - | - |
| Whole Mouse Genome G4122A | A_51_P480602 | NM_009363.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | AK003093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | BC050086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | BC087889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | X51697.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | NM_009363.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | AK003093.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | BC050086.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | BC087889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P480602 | X51697.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115000 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150359 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13123107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13239827 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13239819 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13307495 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13239822 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13239824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31138127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138143 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13172760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13239825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13490259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138133 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13172758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138127 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13394407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13123107 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138128 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31138113 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10449662 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449663 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449665 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10449664 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422448_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10449661 | - | - |
| Mouse Genome 430 2.0 Array | 1422448_at | - | - |
| Mouse Genome 430 2.0 Array | 1422448_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21785_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21785_at | - | - |
| Mouse Expression Set 430 | 1422448_at | - | - |
| Mouse Expression Set 430 | 1422448_PM_at | - | - |
| Murine Genome U74v2 Set | 93302_PM_at | - | - |
| Murine Genome U74v2 Set | 93302_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422448_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422448_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422448_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422448_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 931987 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 940156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940156 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228211 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228211 | - | - |
| mouse-6 v1.1 (Illumina) | scl50084.4.1_68-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940156 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50084.4.1_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228211 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228211 | - | - |

Tff2
Genomic Map