Assay Details
| Gene Symbol: Npy1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK162898.1 | - | 3 - 4 | 553 | 138 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuropeptide Y receptor Y5 | Npy5r |
NPY5-R, NPYY5-R, Y5R |
-12.8kb | Anti-sense | Chr. 8 - 66679965 - 66688581 on NCBI Build GRCm38 |
| Downstream | nuclear assembly factor 1 homolog (S. cerevisiae) | Naf1 | Gm174 | 156.0kb | Sense | Chr. 8 - 66859957 - 66890564 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Npy1r
Genomic Map