Assay Details
| Gene Symbol: Tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008495.1 | - | 1 - 2 | 100 | 95 | |
| AK089021.1 | - | 1 - 2 | 99 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK008495.1, AK089021.1 transcripts.
Set Membership: |
|

Tmem14c
Genomic Map