Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045675.1 | - | 2 - 3 | 275 | 78 | |
| AK082652.1 | - | 1 - 2 | 1447 | 78 | ||
| BC059216.1 | - | 1 - 2 | 550 | 78 | ||
| BC080307.1 | - | 2 - 3 | 108 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045675.1, AK082652.1 transcripts.
Set Membership: |
|

Tmem44
Genomic Map