Assay Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019778.2 | NP_062752.2 | 1 - 2 | 132 | 78 | |
| NM_181058.1 | NP_851401.1 | 1 - 2 | 132 | 78 | ||
| GenBank mRNA | AK005028.2 | - | 1 - 2 | 133 | 78 | |
| BC031114.1 | - | 1 - 2 | 76 | 4971586 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005028.2, BC031114.1, NM_019778.2, NM_181058.1 transcripts.
Set Membership: |
|

Zbtb20
Genomic Map