Assay Details
| Gene Symbol: Vipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011703.4 | NP_035833.2 | 7 - 8 | 922 | 114 | |
| GenBank mRNA | AF266282.1 | - | 7 - 8 | 804 | 114 | |
| AK052465.1 | - | 7 - 8 | 923 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vipr1
Genomic Map