Assay Details
| Gene Symbol: Tal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009317.3 | NP_033343.1 | 1 - 2 | 73 | 92 | |
| GenBank mRNA | AK018448.1 | - | 1 - 2 | 74 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018448.1, NM_009317.3 transcripts.
Set Membership: |
|

Tal2
Genomic Map