Assay Details
| Gene Symbol: Trim17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031172.2 | NP_112449.1 | 1 - 2 | 90 | 66 | |
| GenBank mRNA | AK077212.1 | - | 1 - 2 | 71 | 66 | |
| BC055112.1 | - | 1 - 2 | 90 | 6707359 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC055112.1, NM_031172.2 transcripts.
Set Membership: |
|

Trim17
Genomic Map