Assay Details
| Gene Symbol: Trim17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031172.2 | NP_112449.1 | 2 - 3 | 526 | 87 | |
| GenBank mRNA | AK077212.1 | - | 2 - 3 | 507 | 87 | |
| BC055112.1 | - | 2 - 3 | 526 | 6707359 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim17
Genomic Map