Assay Details
| Gene Symbol: Gm13016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043727.1 | - | 1 - 2 | 86 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 33895 |
Gm33895 | -54.2kb | Sense | Chr. 4 - 140323445 - 140326384 on NCBI Build GRCm38 | |
| Downstream | Rho guanine nucleotide exchange factor (GEF) 10-like | Arhgef10l | 2810441C07Rik | 132.3kb | Anti-sense | Chr. 4 - 140514485 - 140665905 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm13016
Genomic Map