Assay Details
| Gene Symbol: Afm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145146.2 | NP_660128.2 | 4 - 5 | 519 | 159 | |
| GenBank mRNA | AJ011080.1 | - | 4 - 5 | 547 | 159 | |
| AK143987.1 | - | 4 - 5 | 520 | 159 | ||
| BC026681.1 | - | 4 - 5 | 437 | 159 | ||
| BC100597.1 | - | 4 - 5 | 519 | 6767654 | 159 | |
More Information
Set Membership: |
|

Afm
Genomic Map