Assay Details
| Gene Symbol: Tmem8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085508.2 | NP_001078977.1 | 9 - 10 | 1689 | 80 | |
| GenBank mRNA | AK029713.1 | - | 9 - 10 | 1690 | 80 | |
| BC147569.1 | - | 9 - 10 | 1560 | 9056559 | 80 | |
| BC147574.1 | - | 9 - 10 | 1560 | 9056564 | 80 | |
| BC151067.1 | - | 8 - 9 | 1291 | 9087980 | 80 | |
| BC151070.1 | - | 9 - 10 | 1560 | 9087983 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem8b
Genomic Map