Assay Details
| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005740.1 | NP_001005740.1 | 8 - 9 | 1619 | 60 | |
| GenBank mRNA | AK048208.1 | - | 8 - 9 | 1515 | 60 | |
| AK139452.1 | - | 8 - 9 | 1591 | 60 | ||
| AY993932.1 | - | 9 - 10 | 1556 | 60 | ||
| BC043065.1 | - | 3 - 4 | 645 | 60 | ||
| BC048407.1 | - | 7 - 8 | 1450 | 60 | ||
| BC061691.1 | - | 8 - 9 | 1619 | 6307669 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phactr1
Genomic Map