Assay Details
| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047654.1 | - | 1 - 2 | 130 | 138 | |
| AY993932.1 | - | 1 - 2 | 116 | 138 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY993932.1 transcripts.
Set Membership: |
|

Phactr1
Genomic Map