Assay Details
| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005740.1 | NP_001005740.1 | 3 - 4 | 519 | 92 | |
| GenBank mRNA | AK048208.1 | - | 3 - 4 | 415 | 92 | |
| AK139452.1 | - | 3 - 4 | 491 | 92 | ||
| AY993932.1 | - | 5 - 6 | 663 | 92 | ||
| BC048407.1 | - | 3 - 4 | 557 | 92 | ||
| BC061691.1 | - | 3 - 4 | 519 | 6307669 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phactr1
Genomic Map