Assay Details
| Gene Symbol: Phactr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005740.1 | NP_001005740.1 | 6 - 7 | 1214 | 72 | |
| GenBank mRNA | AK048208.1 | - | 6 - 7 | 1110 | 72 | |
| AK139452.1 | - | 6 - 7 | 1186 | 72 | ||
| AY993932.1 | - | 7 - 8 | 1151 | 72 | ||
| BC043065.1 | - | 1 - 2 | 240 | 72 | ||
| BC048407.1 | - | 5 - 6 | 1045 | 72 | ||
| BC061691.1 | - | 6 - 7 | 1214 | 6307669 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phactr1
Genomic Map