| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900027208.mm.1 |
NM_001037749.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027208.mm.1 |
NM_001037749.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027208.mm.1 |
TC0900003235.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027203.mm.1 |
NM_001037749.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027203.mm.1 |
NM_001037749.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027203.mm.1 |
TC0900003235.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027204.mm.1 |
NM_001037749.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027204.mm.1 |
NM_001037749.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027204.mm.1 |
TC0900003235.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027206.mm.1 |
NM_001037749.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027206.mm.1 |
NM_001037749.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027206.mm.1 |
TC0900003235.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900027202.mm.1 |
NM_001037749.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027202.mm.1 |
NM_001037749.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900027202.mm.1 |
TC0900003235.mm.1 |
MTA TC |