Assay Details
| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037749.2 | NP_001032838.1 | 4 - 5 | 740 | 62 | |
| GenBank mRNA | AK133291.1 | - | 4 - 5 | 754 | 62 | |
| BC100471.1 | - | 4 - 5 | 740 | 30917628 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a14
Genomic Map