Assay Details
| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037749.2 | NP_001032838.1 | 10 - 11 | 1605 | 95 | |
| GenBank mRNA | AK133291.1 | - | 10 - 11 | 1618 | 95 | |
| BC100471.1 | - | 10 - 11 | 1605 | 30917628 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a14
Genomic Map