Assay Details
| Gene Symbol: Slc22a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037749.2 | NP_001032838.1 | 11 - 12 | 1807 | 81 | |
| GenBank mRNA | AK133291.1 | - | 11 - 12 | 1820 | 81 | |
| BC100471.1 | - | 11 - 12 | 1807 | 30917628 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P678393 | NM_001037749.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P678393 | AK133291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P678393 | BC100471.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P678393 | NM_001037749.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P678393 | AK133291.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P678393 | BC100471.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 529589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411360 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820198 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 602519 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294469 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445946 | AK034833.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445946 | AK138658.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186658 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23975 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47366 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P678393 | NM_001037749.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P678393 | AK133291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P678393 | BC100471.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P678393 | NM_001037749.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P678393 | AK133291.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P678393 | BC100471.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871043 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26871033 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06946145 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02043943 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06946130 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02043951 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02043977 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06946158 | - | - |
| Whole Mouse Genome G4122A | A_52_P678393 | NM_001037749.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P678393 | AK133291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P678393 | BC100471.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P678393 | NM_001037749.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P678393 | AK133291.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P678393 | BC100471.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02043977 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02043943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26871043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26870930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07489399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07700995 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08096013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871045 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07489400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07700997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871043 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26871051 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10597601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597611 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597608 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597599 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10597607 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1440058_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10597598 | - | - |
| Mouse Genome 430 2.0 Array | 1440058_at | - | - |
| Mouse Genome 430 2.0 Array | 1440058_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382113_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 382113_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900027192.mm.1 | NM_001037749.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027192.mm.1 | NM_001037749.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027192.mm.1 | TC0900003235.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900027193.mm.1 | NM_001037749.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027193.mm.1 | NM_001037749.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900027193.mm.1 | TC0900003235.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1440058_at | - | - |
| Mouse Expression Set 430 | 1440058_PM_at | - | - |
| Murine Genome U74v2 Set | 139397_at | - | - |
| Murine Genome U74v2 Set | 139397_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440058_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1440058_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 310231 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2874193 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2874193 | - | - |

Slc22a14
Genomic Map