Assay Details
| Gene Symbol: Mrpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175001.3 | NP_778166.2 | 6 - 7 | 436 | 95 | |
| GenBank mRNA | AK077615.1 | - | 6 - 7 | 430 | 95 | |
| AK086913.1 | - | 6 - 7 | 415 | 95 | ||
| AK164417.1 | - | 6 - 7 | 415 | 95 | ||
| BC061191.1 | - | 6 - 7 | 426 | 6703228 | 95 | |
More Information
Set Membership: |
|

Mrpl22
Genomic Map