Assay Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081188.1 | NP_001074657.1 | 1 - 2 | 92 | 62 | |
| GenBank mRNA | AK011294.1 | - | 1 - 2 | 79 | 62 | |
| AK129057.1 | - | 1 - 2 | 44 | 62 | ||
| AK217612.1 | - | 175 | 62 | |||
| BC039655.1 | - | 1 - 2 | 77 | 62 | ||
| BC052656.1 | - | 1 - 2 | 74 | 30041585 | 62 | |
| BC094932.1 | - | 1 - 2 | 85 | 30008071 | 62 | |
| BC132363.1 | - | 1 - 2 | 107 | 40130640 | 62 | |
| BC145675.1 | - | 1 - 2 | 107 | 40130978 | 62 | |
More Information
Set Membership: |
|

Exosc7
Genomic Map