| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00011264.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011264.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011264.mm.1 |
TC0X00001782.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00011265.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011265.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011265.mm.1 |
TC0X00001782.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00011266.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011266.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011266.mm.1 |
TC0X00001782.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00011272.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011272.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011272.mm.1 |
TC0X00001782.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00011273.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011273.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011273.mm.1 |
TC0X00001782.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00011274.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011274.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011274.mm.1 |
TC0X00001782.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00011283.mm.1 |
NM_207202.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011283.mm.1 |
NM_207202.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00011283.mm.1 |
TC0X00001782.mm.1 |
MTA TC |