Assay Details
| Gene Symbol: Ccdc120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207202.2 | NP_997085.2 | 4 - 5 | 491 | 73 | |
| GenBank mRNA | BC053918.1 | - | 4 - 5 | 454 | 73 | |
| BC139064.1 | - | 3 - 4 | 267 | 8862086 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc120
Genomic Map