Assay Details
| Gene Symbol: Ccdc120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207202.2 | NP_997085.2 | 6 - 7 | 914 | 58 | |
| GenBank mRNA | BC139064.1 | - | 5 - 6 | 690 | 8862086 | 58 |
| BU505792.1 | - | 1 - 2 | 124 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc120
Genomic Map