Assay Details
| Gene Symbol: Ccdc120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207202.2 | NP_997085.2 | 7 - 8 | 982 | 73 | |
| GenBank mRNA | BC053918.1 | - | 7 - 8 | 947 | 73 | |
| BC139064.1 | - | 6 - 7 | 758 | 8862086 | 73 | |
| BU505792.1 | - | 2 - 3 | 192 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc120
Genomic Map