Assay Details
| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080438.2 | NP_536686.2 | 5 - 6 | 653 | 116 | |
| GenBank mRNA | AF362764.1 | - | 5 - 6 | 571 | 116 | |
| AY230204.1 | - | 5 - 6 | 592 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glra3
Genomic Map