Assay Details
| Gene Symbol: Phf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011078.3 | NP_035208.2 | 14 - 15 | 2045 | 103 | |
| GenBank mRNA | AB221590.1 | - | 3 - 4 | 431 | 103 | |
| AF043726.1 | - | 14 - 15 | 2045 | 103 | ||
| AK172995.1 | - | 12 - 13 | 1752 | 103 | ||
| BC051633.1 | - | 14 - 15 | 2043 | 6518591 | 103 | |
More Information
Set Membership: |
|

Phf2
Genomic Map