Assay Details
| Gene Symbol: E130308A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015681.2 | NP_001015681.1 | 3 - 4 | 1646 | 70 | |
| GenBank mRNA | AK047478.1 | - | 3 - 4 | 1647 | 70 | |
| AK081604.1 | - | 3 - 4 | 1530 | 70 | ||
| AK087509.1 | - | 3 - 4 | 1643 | 70 | ||
| BC017649.1 | - | 3 - 4 | 1413 | 4924105 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

E130308A19Rik
Genomic Map