Assay Details
| Gene Symbol: Wnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175638.3 | NP_783569.1 | 2 - 3 | 883 | 94 | |
| GenBank mRNA | AY184228.1 | - | 2 - 3 | 852 | 94 | |
| AY187027.1 | - | 2 - 3 | 808 | 94 | ||
| AY260059.1 | - | 2 - 3 | 846 | 94 | ||
| BC043677.1 | - | 2 - 3 | 256 | 94 | ||
| BC096453.1 | - | 2 - 3 | 839 | 4234311 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY184228.1 transcripts.
Set Membership: |
|

Wnk4
Genomic Map