Assay Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008112.4 | NP_032138.3 | 1 - 2 | 267 | 92 | |
| GenBank mRNA | AK007871.1 | - | 1 - 2 | 210 | 92 | |
| AK013104.1 | - | 1 - 2 | 212 | 92 | ||
| AK078144.1 | - | 210 | 92 | |||
| AK078514.1 | - | 1 - 2 | 214 | 92 | ||
| AK083063.1 | - | 1 - 2 | 208 | 92 | ||
| AK090083.1 | - | 1 - 2 | 215 | 92 | ||
| AK137761.1 | - | 1 - 2 | 214 | 92 | ||
| AK143633.1 | - | 1 - 2 | 212 | 92 | ||
| AK143681.1 | - | 1 - 2 | 246 | 92 | ||
| AK149958.1 | - | 1 - 2 | 220 | 92 | ||
| AK150356.1 | - | 1 - 2 | 201 | 92 | ||
| AK150655.1 | - | 1 - 2 | 237 | 92 | ||
| AK152250.1 | - | 1 - 2 | 201 | 92 | ||
| AK153381.1 | - | 1 - 2 | 180 | 92 | ||
| AK159734.1 | - | 1 - 2 | 212 | 92 | ||
| AK166317.1 | - | 1 - 2 | 212 | 92 | ||
| AK167659.1 | - | 1 - 2 | 224 | 92 | ||
| AK169299.1 | - | 1 - 2 | 178 | 92 | ||
| BC024971.1 | - | 1 - 2 | 188 | 92 | ||
| BC053381.1 | - | 2367 | 92 | |||
| BC055341.1 | - | 1 - 2 | 193 | 6808475 | 92 | |
| BY072982.1 | - | 1 - 2 | 280 | 92 | ||
| L36314.1 | - | 1 - 2 | 73 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007871.1,AK013104.1,AK078144.1,AK090083.1,AK137761.1,AK143633.1,AK149958.1,AK152250.1,AK153381.1,AK159734.1,AK166317.1,AK167659.1,AK169299.1,BC055341.1,L36314.1,NM_008112.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdi2
Genomic Map