Assay Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008112.4 | NP_032138.3 | 2 - 3 | 373 | 71 | |
| GenBank mRNA | AK007871.1 | - | 2 - 3 | 316 | 71 | |
| AK013104.1 | - | 2 - 3 | 318 | 71 | ||
| AK078144.1 | - | 316 | 71 | |||
| AK078514.1 | - | 2 - 3 | 320 | 71 | ||
| AK083063.1 | - | 2 - 3 | 314 | 71 | ||
| AK090083.1 | - | 2 - 3 | 321 | 71 | ||
| AK137761.1 | - | 2 - 3 | 320 | 71 | ||
| AK143633.1 | - | 2 - 3 | 318 | 71 | ||
| AK143681.1 | - | 2 - 3 | 352 | 71 | ||
| AK149958.1 | - | 2 - 3 | 326 | 71 | ||
| AK150356.1 | - | 2 - 3 | 307 | 71 | ||
| AK150655.1 | - | 2 - 3 | 343 | 71 | ||
| AK152250.1 | - | 2 - 3 | 307 | 71 | ||
| AK153381.1 | - | 2 - 3 | 286 | 71 | ||
| AK159734.1 | - | 2 - 3 | 318 | 71 | ||
| AK166317.1 | - | 2 - 3 | 318 | 71 | ||
| AK167659.1 | - | 2 - 3 | 330 | 71 | ||
| AK169299.1 | - | 2 - 3 | 284 | 71 | ||
| BC024971.1 | - | 2 - 3 | 294 | 71 | ||
| BC053381.1 | - | 2473 | 71 | |||
| BC055341.1 | - | 2 - 3 | 299 | 6808475 | 71 | |
| L36314.1 | - | 2 - 3 | 179 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdi2
Genomic Map