Assay Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008112.4 | NP_032138.3 | 4 - 5 | 610 | 115 | |
| GenBank mRNA | AK007871.1 | - | 4 - 5 | 553 | 115 | |
| AK013104.1 | - | 4 - 5 | 555 | 115 | ||
| AK078144.1 | - | 553 | 115 | |||
| AK078514.1 | - | 4 - 5 | 557 | 115 | ||
| AK079690.1 | - | 3 - 4 | 451 | 115 | ||
| AK083063.1 | - | 4 - 5 | 552 | 115 | ||
| AK090083.1 | - | 4 - 5 | 558 | 115 | ||
| AK137761.1 | - | 4 - 5 | 557 | 115 | ||
| AK143633.1 | - | 4 - 5 | 555 | 115 | ||
| AK143681.1 | - | 4 - 5 | 589 | 115 | ||
| AK149958.1 | - | 4 - 5 | 563 | 115 | ||
| AK150356.1 | - | 4 - 5 | 545 | 115 | ||
| AK150655.1 | - | 4 - 5 | 580 | 115 | ||
| AK152250.1 | - | 4 - 5 | 544 | 115 | ||
| AK153381.1 | - | 4 - 5 | 523 | 115 | ||
| AK159734.1 | - | 4 - 5 | 555 | 115 | ||
| AK166317.1 | - | 4 - 5 | 555 | 115 | ||
| AK167659.1 | - | 4 - 5 | 567 | 115 | ||
| AK169299.1 | - | 4 - 5 | 521 | 115 | ||
| AK207166.1 | - | 1 - 2 | 100 | 115 | ||
| AK213780.1 | - | 1 - 2 | 99 | 115 | ||
| BC024971.1 | - | 4 - 5 | 531 | 115 | ||
| BC053381.1 | - | 2710 | 115 | |||
| BC055341.1 | - | 4 - 5 | 536 | 6808475 | 115 | |
| L36314.1 | - | 4 - 5 | 416 | 115 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdi2
Genomic Map