Assay Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008112.4 | NP_032138.3 | 7 - 8 | 1049 | 119 | |
| GenBank mRNA | AK078144.1 | - | 992 | 119 | ||
| AK079690.1 | - | 6 - 7 | 890 | 119 | ||
| AK083063.1 | - | 7 - 8 | 991 | 119 | ||
| AK090083.1 | - | 7 - 8 | 997 | 119 | ||
| AK137761.1 | - | 7 - 8 | 996 | 119 | ||
| AK143633.1 | - | 7 - 8 | 994 | 119 | ||
| AK143681.1 | - | 7 - 8 | 1028 | 119 | ||
| AK149958.1 | - | 7 - 8 | 1002 | 119 | ||
| AK150356.1 | - | 7 - 8 | 984 | 119 | ||
| AK150655.1 | - | 7 - 8 | 1019 | 119 | ||
| AK152250.1 | - | 7 - 8 | 983 | 119 | ||
| AK153381.1 | - | 7 - 8 | 962 | 119 | ||
| AK159734.1 | - | 7 - 8 | 994 | 119 | ||
| AK166317.1 | - | 7 - 8 | 994 | 119 | ||
| AK169299.1 | - | 7 - 8 | 960 | 119 | ||
| AK214533.1 | - | 2 - 3 | 122 | 119 | ||
| BC024971.1 | - | 7 - 8 | 970 | 119 | ||
| BC053381.1 | - | 3149 | 119 | |||
| BC055341.1 | - | 7 - 8 | 975 | 6808475 | 119 | |
| L36314.1 | - | 7 - 8 | 855 | 119 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdi2
Genomic Map