Assay Details
| Gene Symbol: Gdi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008112.4 | NP_032138.3 | 8 - 9 | 1211 | 127 | |
| GenBank mRNA | AK007871.1 | - | 8 - 9 | 1154 | 127 | |
| AK078144.1 | - | 1154 | 127 | |||
| AK079690.1 | - | 7 - 8 | 1052 | 127 | ||
| AK083063.1 | - | 8 - 9 | 1153 | 127 | ||
| AK090083.1 | - | 8 - 9 | 1159 | 127 | ||
| AK137761.1 | - | 8 - 9 | 1158 | 127 | ||
| AK143633.1 | - | 8 - 9 | 1156 | 127 | ||
| AK143681.1 | - | 8 - 9 | 1190 | 127 | ||
| AK149958.1 | - | 8 - 9 | 1164 | 127 | ||
| AK150356.1 | - | 8 - 9 | 1146 | 127 | ||
| AK150655.1 | - | 8 - 9 | 1181 | 127 | ||
| AK152250.1 | - | 8 - 9 | 1145 | 127 | ||
| AK153381.1 | - | 8 - 9 | 1124 | 127 | ||
| AK159734.1 | - | 8 - 9 | 1156 | 127 | ||
| AK166317.1 | - | 8 - 9 | 1156 | 127 | ||
| AK167659.1 | - | 8 - 9 | 1168 | 127 | ||
| AK169299.1 | - | 8 - 9 | 1122 | 127 | ||
| BC024971.1 | - | 8 - 9 | 1132 | 127 | ||
| BC053381.1 | - | 3311 | 127 | |||
| BC055341.1 | - | 8 - 9 | 1137 | 6808475 | 127 | |
| L36314.1 | - | 8 - 9 | 1017 | 127 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gdi2
Genomic Map